Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hexyn-3-ol, 95%, Thermo Scientific™

Catalog No. AAL0972706
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0972706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0972706 Supplier Thermo Scientific Chemicals Supplier No. L0972706

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20739-59-7
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.15
MDL Number MFCD00041591
InChI Key KARLLBDFLHNKBO-UHFFFAOYNA-N
Synonym 4-hexyn-3-ol, ethyl 1-propynyl carbinol, 1-pentyn-3-ol, methyl, 4-hexyne-3-ol, acmc-1cpld
PubChem CID 140751
IUPAC Name hex-4-yn-3-ol
SMILES CCC(O)C#CC

Specifications

Density 0.894
Boiling Point 94°C to 95°C (105 mmHg)
Refractive Index 1.45
Quantity 5 g
UN Number UN1987
Beilstein 1739392
Formula Weight 98.15
Percent Purity 95%
Chemical Name or Material 4-Hexyn-3-ol
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.