Learn More
AdipoGen anti-4-Hydroxy-2-nonenal [4-HNE], mAb (HNEJ-2) (100 µg)
SDP
Supplier:  AdipoGen JAIMHN100P
Encompass_Preferred
anti-4-Hydroxy-2-nonenal [4-HNE], mAb (HNEJ-2) (100 µg), AdipoGen Life Sciences. Monoclonal Antibody. Recognizes 4-HNE-lysine, 4-HNE-histidine and 4-HNE cysteine adduct. Has much higher affinity for the 4-HNE-histidine adduct than 4-HNE-lysine or 4-HNE cysteine adduct. Shows almost negligible reactivity with proteins that were treated with other aldehydes, such as 2-nonenal, 2-hexenal, 1-hexanal, 4-hydroxy-2-hexenal, formaldehyde, or glutaraldehyde. Lyophilized. Contains 50mM Tris-buffered saline (TBS). 4-HNE formed inside living body is reported to originate from phospholipid-bound arachidonic acid. 4-HNE may be one of the most reliable biomarker of lipid peroxidation.
Catalog No. 50-148-9749