Learn More
Medchemexpress LLC HY-117433 5mg Medchemexpress, 4-Hydroperoxy cyclophosphamide CAS:39800-16-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1174335MG
Encompass_Preferred
Medchemexpress, HY-117433 5mg 4-Hydroperoxy cyclophosphamide CAS:39800-16-3 4-Hydroperoxy cyclophosphamide is the active metabolite form of the prodrug Cyclophosphamide. 4-Hydroperoxy cyclophosphamide crosslinks DNA and induces T cell apoptosis independent of death receptor activation, but activates mitochondrial death pathways through production of reactive oxygen species (ROS). 4-Hydroperoxy cyclophosphamide has the potential for lymphomas and autoimmune disorders. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0905