Learn More
Medchemexpress LLC 4-hydroperoxy cyclophosphamide | 39800-16-3 | MFCD01710226 | ≥98.0% | 293.08 | C7H15Cl2N2O4P | 1 MG
SDP
Catalog No. 5000227051
Encompass_Preferred
4-hydroperoxy cyclophosphamide is the active metabolite of cyclophosphamide and functions as a DNA alkylator/crosslinker that induces T-cell apoptosis and produces reactive oxygen species. It is used as a research reagent to study DNA damage, apoptosis pathways, and immune modulation in vitro and in vivo.
  • Active DNA crosslinker inducing apoptosis independent of death-receptor activation.
  • Useful for studying apoptosis, immune cell depletion, and autoimmune disease models.
  • Supplied in small mass formats suitable for stock solution preparation, including 1 mg quantities.
  • Stable when stored under nitrogen at -20°C; in solvent stable at -80°C for up to 6 months.
  • High purity (≥98.0%) suitable for biochemical and cellular assays.
Catalog No. 50-002-27051 Supplier Medchemexpress LLC Supplier No. HY1174331MG
4-hydroperoxy cyclophosphamide is the active metabolite of cyclophosphamide and functions as a DNA alkylator/crosslinker that induces T-cell apoptosis and produces reactive oxygen species. It is used as a research reagent to study DNA damage, apoptosis pathways, and immune modulation in vitro and in vivo.
  • Active DNA crosslinker inducing apoptosis independent of death-receptor activation.
  • Useful for studying apoptosis, immune cell depletion, and autoimmune disease models.
  • Supplied in small mass formats suitable for stock solution preparation, including 1 mg quantities.
  • Stable when stored under nitrogen at -20°C; in solvent stable at -80°C for up to 6 months.
  • High purity (≥98.0%) suitable for biochemical and cellular assays.