Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-hydroperoxy cyclophosphamide | 39800-16-3 | MFCD01710226 | ≥98.0% | 293.08 g/mol | C7H15Cl2N2O4P | 100 MG
SDP

Catalog No. 5000227545
Encompass_Preferred

4-Hydroperoxy cyclophosphamide is the hydroperoxide derivative of cyclophosphamide supplied for in vitro research. It acts as a reactive alkylating agent and a source of activated cyclophosphamide metabolites for biochemical and cellular studies. Supplied as a solid, the material requires cold, inert storage and is for research use only, not for human or clinical applications.

  • CAS number: 39800-16-3
  • Molecular weight: 293.08 g/mol
  • Chemical formula: C7H15Cl2N2O4P
  • Purity: ≥98.0% (as stated on COA)
  • Solubility: DMSO 50 mg/mL; ultrasonic assistance recommended
  • Storage: -20°C under nitrogen; in solvent store at -80°C for up to 6 months
  • Intended use: for research use only; not sold to patients

Catalog No. 50-002-27545 Supplier Medchemexpress LLC Supplier No. HY117433100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Hydroperoxy cyclophosphamide is the hydroperoxide derivative of cyclophosphamide supplied for in vitro research. It acts as a reactive alkylating agent and a source of activated cyclophosphamide metabolites for biochemical and cellular studies. Supplied as a solid, the material requires cold, inert storage and is for research use only, not for human or clinical applications.

  • CAS number: 39800-16-3
  • Molecular weight: 293.08 g/mol
  • Chemical formula: C7H15Cl2N2O4P
  • Purity: ≥98.0% (as stated on COA)
  • Solubility: DMSO 50 mg/mL; ultrasonic assistance recommended
  • Storage: -20°C under nitrogen; in solvent store at -80°C for up to 6 months
  • Intended use: for research use only; not sold to patients

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.