Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-hydroperoxy cyclophosphamide | 39800-16-3 | MFCD01710226 | >98.0% | 293.08 g/mol | C7H15Cl2N2O4P | 25 MG
SDP

Catalog No. 5000227052
Encompass_Preferred

4-Hydroperoxy cyclophosphamide is the active metabolite of cyclophosphamide and is used as a research reagent to study DNA crosslinking, cytotoxic mechanisms, and immune-cell apoptosis. It is supplied as a dry solid for laboratory use and requires low-temperature storage to maintain stability in storage and handling.

  • active metabolite used in cytotoxicity and immunology studies.
  • cross-links DNA and induces T-cell apoptosis.
  • high purity: >98.0% (HPLC) as reported by analytical sources.
  • molecular weight 293.08 g/mol and formula C7H15Cl2N2O4P.
  • soluble in DMSO at 50 mg/mL; sonication may be required.
  • store at -20°C under nitrogen; in solvent store at -80°C for long-term stability.

Catalog No. 50-002-27052 Supplier Medchemexpress LLC Supplier No. HY11743325MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Hydroperoxy cyclophosphamide is the active metabolite of cyclophosphamide and is used as a research reagent to study DNA crosslinking, cytotoxic mechanisms, and immune-cell apoptosis. It is supplied as a dry solid for laboratory use and requires low-temperature storage to maintain stability in storage and handling.

  • active metabolite used in cytotoxicity and immunology studies.
  • cross-links DNA and induces T-cell apoptosis.
  • high purity: >98.0% (HPLC) as reported by analytical sources.
  • molecular weight 293.08 g/mol and formula C7H15Cl2N2O4P.
  • soluble in DMSO at 50 mg/mL; sonication may be required.
  • store at -20°C under nitrogen; in solvent store at -80°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.