Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-hydroperoxy cyclophosphamide-d4 | 1246816-71-6 | MFCD34567216 | 95.0% | 297.11 | C7H11D4Cl2N2O4P | 100 UG
SDP

Catalog No. 5000226785
Encompass_Preferred

4-Hydroperoxy cyclophosphamide-d4 is a deuterium-labeled analog of 4-hydroperoxy cyclophosphamide used as an internal standard and tracer in analytical and biochemical research, particularly for mass spectrometry and metabolic studies. It supports precise quantitation and method development for small-scale laboratory experiments.

  • Deuterium-labeled internal standard for MS and metabolic studies.
  • Reported purity 95.0% suitable for analytical applications.
  • Molecular weight 297.11 with formula C7H11D4Cl2N2O4P for exact mass reference.
  • Provided in a 100 UG quantity for small-scale experiments and method development.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-26785 Supplier Medchemexpress LLC Supplier No. HY117433S100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Hydroperoxy cyclophosphamide-d4 is a deuterium-labeled analog of 4-hydroperoxy cyclophosphamide used as an internal standard and tracer in analytical and biochemical research, particularly for mass spectrometry and metabolic studies. It supports precise quantitation and method development for small-scale laboratory experiments.

  • Deuterium-labeled internal standard for MS and metabolic studies.
  • Reported purity 95.0% suitable for analytical applications.
  • Molecular weight 297.11 with formula C7H11D4Cl2N2O4P for exact mass reference.
  • Provided in a 100 UG quantity for small-scale experiments and method development.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.