Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-hydroperoxy cyclophosphamide-d4 | 1246816-71-6 | MFCD34567216 | 95.0% | 297.11 g·mol⁻¹ | C7H11D4Cl2N2O4P | 1 MG
SDP

Catalog No. 5000226251
Encompass_Preferred

4-Hydroperoxy cyclophosphamide-d4 is the deuterium-labeled stable isotope analog of 4-hydroperoxy cyclophosphamide, provided as a solid for analytical and research use in studies of drug metabolism and mechanism. It is intended for use as an internal standard and research reagent in small-quantity analytical workflows.

  • Deuterium-labeled (d4) analog for use as an internal standard in analytical assays.
  • Suitable for studies of cyclophosphamide metabolism and mechanism.
  • Molecular weight 297.11 g·mol⁻¹.
  • Molecular formula C7H11D4Cl2N2O4P.
  • Available in 1 mg, 5 mg, and 10 mg sizes.
  • Supplied as a solid; follow SDS handling instructions for storage and safety.

Catalog No. 50-002-26251 Supplier Medchemexpress LLC Supplier No. HY117433S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Hydroperoxy cyclophosphamide-d4 is the deuterium-labeled stable isotope analog of 4-hydroperoxy cyclophosphamide, provided as a solid for analytical and research use in studies of drug metabolism and mechanism. It is intended for use as an internal standard and research reagent in small-quantity analytical workflows.

  • Deuterium-labeled (d4) analog for use as an internal standard in analytical assays.
  • Suitable for studies of cyclophosphamide metabolism and mechanism.
  • Molecular weight 297.11 g·mol⁻¹.
  • Molecular formula C7H11D4Cl2N2O4P.
  • Available in 1 mg, 5 mg, and 10 mg sizes.
  • Supplied as a solid; follow SDS handling instructions for storage and safety.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.