Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-hydroperoxycyclophosphamide | 39800-16-3 | MFCD01710226 | >98.0% | 293.08 g/mol | C7H15Cl2N2O4P | 50 MG
SDP

Catalog No. 5000227640
Encompass_Preferred

4-hydroperoxycyclophosphamide is the activated hydroperoxide metabolite of cyclophosphamide used as a research reagent to study DNA cross-linking, apoptosis, and reactive oxygen species (ROS)-mediated mitochondrial death pathways. It is supplied for research use only and not for human or clinical use.

  • Activated metabolite that cross-links DNA and induces T cell apoptosis.
  • Useful for in vitro studies of apoptosis, oxidative stress, and DNA damage.
  • High solubility in DMSO (50 mg/mL); ultrasonic agitation recommended for dissolution.
  • Storage guidance: store at -20°C under inert gas; in solution store at -80°C up to 6 months, or -20°C up to 1 month under nitrogen.
  • Molecular weight 293.08 g/mol; formula C7H15Cl2N2O4P.
  • For research use only; not intended for human or clinical applications.

Catalog No. 50-002-27640 Supplier Medchemexpress LLC Supplier No. HY11743350MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-hydroperoxycyclophosphamide is the activated hydroperoxide metabolite of cyclophosphamide used as a research reagent to study DNA cross-linking, apoptosis, and reactive oxygen species (ROS)-mediated mitochondrial death pathways. It is supplied for research use only and not for human or clinical use.

  • Activated metabolite that cross-links DNA and induces T cell apoptosis.
  • Useful for in vitro studies of apoptosis, oxidative stress, and DNA damage.
  • High solubility in DMSO (50 mg/mL); ultrasonic agitation recommended for dissolution.
  • Storage guidance: store at -20°C under inert gas; in solution store at -80°C up to 6 months, or -20°C up to 1 month under nitrogen.
  • Molecular weight 293.08 g/mol; formula C7H15Cl2N2O4P.
  • For research use only; not intended for human or clinical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.