Learn More
Medchemexpress LLC [4-hydroxy-2-(2-hydroxyphenyl)-1,3-thiazol-5-yl]-(1,2-oxazolidin-2-yl)methanone | 2102179-29-1 | MFCD34602961 | 99.5% | 292.31 g/mol | C13H12N2O4S | 5 MG
SDP
Catalog No. 5000221092
Encompass_Preferred
TRPM8 antagonist 3 is a novel, potent small-molecule blocker of the TRPM8 ion channel with a reported IC50 of 11 nM. Supplied as a solid (off-white to light yellow), it is intended for research use and is characterized by high purity and defined storage conditions.
  • Potent TRPM8 inhibition (IC50 11 nM).
  • High purity (≈99.5%).
  • Chemical formula C13H12N2O4S; molecular weight 292.31 g/mol.
  • Supplied as an off-white to light yellow solid.
  • Powder stable at -20°C for up to 3 years; solvent stability documented.
  • Available in multiple package sizes and as a 10 mM solution in DMSO.
Catalog No. 50-002-21092 Supplier Medchemexpress LLC Supplier No. HY1451245MG
TRPM8 antagonist 3 is a novel, potent small-molecule blocker of the TRPM8 ion channel with a reported IC50 of 11 nM. Supplied as a solid (off-white to light yellow), it is intended for research use and is characterized by high purity and defined storage conditions.
  • Potent TRPM8 inhibition (IC50 11 nM).
  • High purity (≈99.5%).
  • Chemical formula C13H12N2O4S; molecular weight 292.31 g/mol.
  • Supplied as an off-white to light yellow solid.
  • Powder stable at -20°C for up to 3 years; solvent stability documented.
  • Available in multiple package sizes and as a 10 mM solution in DMSO.