Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4'-Hydroxy-2,4-dimethoxychalcone | 151135-64-7 | 5 MG
SDP

Catalog No. 5000419032
Encompass_Preferred

4'-Hydroxy-2,4-dimethoxychalcone is a natural chalcone derivative found in the red herbal resin of *Dracaena cochinchinensis*. It exhibits a wide range of biological activities, including antimalarial, antiprotozoal, antibacterial, and antifungal properties. This product is intended for research use only.

  • Purity of 99.76%
  • Exhibits antimalarial, antiprotozoal, antibacterial, and antifungal activity
  • Ships at room temperature in continental US
  • Store at -20°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light
  • Soluble in DMSO at 50 mg/mL
  • Active against *P. falciparum* D6 (IC50=4.2 μM) and W2 (IC50=4.8 μM)

Catalog No. 50-004-19032 Supplier Medchemexpress LLC Supplier No. HYN75165MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4'-Hydroxy-2,4-dimethoxychalcone is a natural chalcone derivative found in the red herbal resin of *Dracaena cochinchinensis*. It exhibits a wide range of biological activities, including antimalarial, antiprotozoal, antibacterial, and antifungal properties. This product is intended for research use only.

  • Purity of 99.76%
  • Exhibits antimalarial, antiprotozoal, antibacterial, and antifungal activity
  • Ships at room temperature in continental US
  • Store at -20°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light
  • Soluble in DMSO at 50 mg/mL
  • Active against *P. falciparum* D6 (IC50=4.2 μM) and W2 (IC50=4.8 μM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.