Learn More
Medchemexpress LLC 4-hydroxy-2-butanone | 590-90-9 | 98.2% | 88.11 g/mol | C4H8O2 | 50 G
SDP
Catalog No. 5000210118
Encompass_Preferred
4-hydroxy-2-butanone (CAS 590-90-9) is a beta-hydroxy ketone identified as a free volatile component of glycoside precursors in wax gourd. It is supplied for research use and is used as a synthetic building block and intermediate in chemical and biochemical studies. The product is provided as a solid with reported purity of 98.2%.
  • Free volatile component of glycoside precursors in wax gourd.
  • Reported purity 98.2%.
  • Molecular formula C4H8O2; molecular weight 88.11 g/mol.
  • Available in solid pack sizes including 50 G and solution formats (10 mM in DMSO).
  • Intended for research use and not for human or clinical use.
Catalog No. 50-002-10118 Supplier Medchemexpress LLC Supplier No. HYW05128750G
4-hydroxy-2-butanone (CAS 590-90-9) is a beta-hydroxy ketone identified as a free volatile component of glycoside precursors in wax gourd. It is supplied for research use and is used as a synthetic building block and intermediate in chemical and biochemical studies. The product is provided as a solid with reported purity of 98.2%.
  • Free volatile component of glycoside precursors in wax gourd.
  • Reported purity 98.2%.
  • Molecular formula C4H8O2; molecular weight 88.11 g/mol.
  • Available in solid pack sizes including 50 G and solution formats (10 mM in DMSO).
  • Intended for research use and not for human or clinical use.