Learn More
Medchemexpress LLC 4-hydroxy-2-butanone | 590-90-9 | MFCD00059005 | 98.2% | 88.11 g/mol | C4H8O2 | 100 G
SDP
Catalog No. 5000210298
Encompass_Preferred
4-hydroxy-2-butanone (CAS 590-90-9) is a beta-hydroxy ketone and volatile component of glycoside precursors in wax gourd. It is supplied as a research reagent and is used as a synthetic intermediate and analytical standard in organic and analytical chemistry.
  • High purity, 98.2% by GC.
  • Chemical formula C4H8O2 and molecular weight 88.11 g/mol.
  • Available in multiple package sizes, including 100 g.
  • Stable at room temperature for up to 3 years when stored properly.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-10298 Supplier Medchemexpress LLC Supplier No. HYW051287100G
4-hydroxy-2-butanone (CAS 590-90-9) is a beta-hydroxy ketone and volatile component of glycoside precursors in wax gourd. It is supplied as a research reagent and is used as a synthetic intermediate and analytical standard in organic and analytical chemistry.
  • High purity, 98.2% by GC.
  • Chemical formula C4H8O2 and molecular weight 88.11 g/mol.
  • Available in multiple package sizes, including 100 g.
  • Stable at room temperature for up to 3 years when stored properly.
  • Intended for research use only; not for human or clinical use.