Learn More
Medchemexpress LLC 4-hydroxy-2-butanone | 590-90-9 | MFCD00059005 | 98.2% | 88.11 g/mol | C4H8O2 | 25 G
SDP
Catalog No. 5000209654
Encompass_Preferred
4-Hydroxy-2-butanone is a volatile beta-hydroxy ketone used as a research reagent and as a component of glycoside precursors in plant metabolite studies. It is supplied in solution or bulk liquid formats for laboratory use; this product is intended for research use only and not for human consumption.
  • High purity: 98.2%.
  • Molecular formula C4H8O2 and molecular weight 88.11 g/mol.
  • Available in multiple pack sizes, including 25 G.
  • Provided as a liquid; may be supplied in solution or neat depending on packaging.
  • Suitable as a synthetic intermediate and for metabolite or glycoside precursor research.
Catalog No. 50-002-09654 Supplier Medchemexpress LLC Supplier No. 25GHYW051287
4-Hydroxy-2-butanone is a volatile beta-hydroxy ketone used as a research reagent and as a component of glycoside precursors in plant metabolite studies. It is supplied in solution or bulk liquid formats for laboratory use; this product is intended for research use only and not for human consumption.
  • High purity: 98.2%.
  • Molecular formula C4H8O2 and molecular weight 88.11 g/mol.
  • Available in multiple pack sizes, including 25 G.
  • Provided as a liquid; may be supplied in solution or neat depending on packaging.
  • Suitable as a synthetic intermediate and for metabolite or glycoside precursor research.