Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-hydroxy-2-butanone | 590-90-9 | MFCD00059005 | 98.2% | 88.11 g/mol | C4H8O2 | 5 G
SDP

Catalog No. 5000209830
Encompass_Preferred

4-hydroxy-2-butanone is a beta-hydroxy ketone and a volatile component of glycoside precursors found in wax gourd; it is used as a reagent in laboratory research.

  • CAS number: 590-90-9.
  • Molecular formula: C4H8O2.
  • Molecular weight: 88.11 g/mol.
  • Purity: 98.2%.
  • Suitable for research use and laboratory applications.
  • Available in small pack sizes including 5 G.

Catalog No. 50-002-09830 Supplier Medchemexpress LLC Supplier No. HYW0512875G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-hydroxy-2-butanone is a beta-hydroxy ketone and a volatile component of glycoside precursors found in wax gourd; it is used as a reagent in laboratory research.

  • CAS number: 590-90-9.
  • Molecular formula: C4H8O2.
  • Molecular weight: 88.11 g/mol.
  • Purity: 98.2%.
  • Suitable for research use and laboratory applications.
  • Available in small pack sizes including 5 G.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.