Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-hydroxy-2-butanone | 590-90-9 | MFCD00059005 | 98.2% | 88.11 g/mol | C4H8O2 | 500 MG
SDP

Catalog No. 5000209827
Encompass_Preferred

4-Hydroxy-2-butanone is a small organic beta-hydroxy ketone and a free volatile component of glycoside precursors in wax gourd. This research reagent is supplied in multiple pack sizes for laboratory use and is intended for research use only.

  • Purity 98.2%.
  • Cas number 590-90-9.
  • Molecular formula C4H8O2, molecular weight 88.11 g/mol.
  • Available pack sizes include 10 mM (1 mL in DMSO), 500 mg, 1 g, 5 g, 10 g, 25 g, 50 g, 100 g, and bulk (>100 g).
  • Intended for laboratory research use; not for human or veterinary use.

Catalog No. 50-002-09827 Supplier Medchemexpress LLC Supplier No. HYW051287500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Hydroxy-2-butanone is a small organic beta-hydroxy ketone and a free volatile component of glycoside precursors in wax gourd. This research reagent is supplied in multiple pack sizes for laboratory use and is intended for research use only.

  • Purity 98.2%.
  • Cas number 590-90-9.
  • Molecular formula C4H8O2, molecular weight 88.11 g/mol.
  • Available pack sizes include 10 mM (1 mL in DMSO), 500 mg, 1 g, 5 g, 10 g, 25 g, 50 g, 100 g, and bulk (>100 g).
  • Intended for laboratory research use; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.