Learn More
Medchemexpress LLC 4-hydroxy-2-methyl-1,1-dioxo-N-(1,3-thiazol-2-yl)-1λ6,2-benzothiazine-3-carboxamide | 34042-85-8 | MFCD00866053 | 100.0% | 337.37 g·mol⁻1 | C13H11N3O4S2 | 10 MG
SDP
Catalog No. 5000242107
Encompass_Preferred
Sudoxicam is a non-steroidal anti-inflammatory drug (NSAID) of the enol-carboxamide class that acts as a reversible, orally active cyclooxygenase (COX) antagonist. Provided as a research-grade chemical for in vitro and analytical use.
  • Reversible COX antagonist for inflammation research.
  • Analytical-grade solid with molecular formula C13H11N3O4S2 and molecular weight 337.37 g·mol⁻¹.
  • High purity (99.96%), suitable for research and analytical applications.
  • Available in small package sizes (1 mg, 5 mg, 10 mg) for assay and method development.
  • Storage: powder - -20°C (3 years), 4°C (2 years); in solvent - -80°C (6 months), -20°C (1 month).
Catalog No. 50-002-42107 Supplier Medchemexpress LLC Supplier No. HY10662810MG
Sudoxicam is a non-steroidal anti-inflammatory drug (NSAID) of the enol-carboxamide class that acts as a reversible, orally active cyclooxygenase (COX) antagonist. Provided as a research-grade chemical for in vitro and analytical use.
  • Reversible COX antagonist for inflammation research.
  • Analytical-grade solid with molecular formula C13H11N3O4S2 and molecular weight 337.37 g·mol⁻¹.
  • High purity (99.96%), suitable for research and analytical applications.
  • Available in small package sizes (1 mg, 5 mg, 10 mg) for assay and method development.
  • Storage: powder - -20°C (3 years), 4°C (2 years); in solvent - -80°C (6 months), -20°C (1 month).