Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-hydroxy-2-methyl-1,1-dioxo-N-(1,3-thiazol-2-yl)-1λ6,2-benzothiazine-3-carboxamide | 34042-85-8 | MFCD00866053 | 100.0% | 337.37 g/mol | C13H11N3O4S2 | 100 MG
SDP

Catalog No. 5000241425
Encompass_Preferred

Sudoxicam is an enol-carboxamide non-steroidal anti-inflammatory compound that functions as a reversible cyclooxygenase (COX) antagonist. It is provided as a high-purity analytical reagent intended for in-vitro and preclinical research use only (not for human or clinical use).

  • Purity: 99.96%.
  • CAS number: 34042-85-8.
  • Molecular weight: 337.37 g/mol.
  • Molecular formula: C13H11N3O4S2.
  • Typical pack size: 100 mg solid.
  • Intended use: research reagent and analytical standard for COX inhibition studies.

Catalog No. 50-002-41425 Supplier Medchemexpress LLC Supplier No. HY106628100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sudoxicam is an enol-carboxamide non-steroidal anti-inflammatory compound that functions as a reversible cyclooxygenase (COX) antagonist. It is provided as a high-purity analytical reagent intended for in-vitro and preclinical research use only (not for human or clinical use).

  • Purity: 99.96%.
  • CAS number: 34042-85-8.
  • Molecular weight: 337.37 g/mol.
  • Molecular formula: C13H11N3O4S2.
  • Typical pack size: 100 mg solid.
  • Intended use: research reagent and analytical standard for COX inhibition studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.