Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-hydroxy-2-methyl-1,1-dioxo-N-(1,3-thiazol-2-yl)-1λ6,2-benzothiazine-3-carboxamide | 34042-85-8 | MFCD00866053 | 100.0% | 337.37 g/mol | C13H11N3O4S2 | 25 MG
SDP

Catalog No. 5000240165
Encompass_Preferred

Sudoxicam is a reversible, orally active cyclooxygenase (COX) antagonist and a non-steroidal anti-inflammatory compound of the enol-carboxamide class supplied for research and analytical use only. It demonstrates anti-inflammatory, anti-edema, and antipyretic activity and is provided as a solid and in DMSO solution formulations for laboratory studies.

  • Reversible, orally active COX antagonist.
  • High reported purity suitable for analytical work.
  • Chemical formula C13H11N3O4S2 and molecular weight 337.37 g/mol.
  • Soluble in DMSO; ultrasonic warming may improve solubility.
  • Stable as powder at -20°C for long-term storage; in solvent storage guidelines provided.

Catalog No. 50-002-40165 Supplier Medchemexpress LLC Supplier No. HY10662825MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sudoxicam is a reversible, orally active cyclooxygenase (COX) antagonist and a non-steroidal anti-inflammatory compound of the enol-carboxamide class supplied for research and analytical use only. It demonstrates anti-inflammatory, anti-edema, and antipyretic activity and is provided as a solid and in DMSO solution formulations for laboratory studies.

  • Reversible, orally active COX antagonist.
  • High reported purity suitable for analytical work.
  • Chemical formula C13H11N3O4S2 and molecular weight 337.37 g/mol.
  • Soluble in DMSO; ultrasonic warming may improve solubility.
  • Stable as powder at -20°C for long-term storage; in solvent storage guidelines provided.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.