Learn More
Medchemexpress LLC 4-hydroxy-2-methyl-1,1-dioxo-N-(1,3-thiazol-2-yl)-1lambda6,2-benzothiazine-3-carboxamide | 34042-85-8 | MFCD00866053 | 95.0% | 337.37 g/mol | C13H11N3O4S2 | 5 MG
SDP
Catalog No. 5000240704
Encompass_Preferred
Sudoxicam is a reversible, orally active cyclooxygenase (COX) antagonist from the enol-carboxamide class of non-steroidal anti-inflammatory drugs (NSAIDs). Provided for laboratory research use only, it exhibits anti-inflammatory, anti-edema, and antipyretic activities and is supplied as a characterized solid for use in in vitro and in vivo pharmacology studies.
  • Reversible COX antagonist with anti-inflammatory, anti-edema, and antipyretic activity.
  • Enol-carboxamide class NSAID commonly used in pharmacology research.
  • Supplied as a solid suitable for dissolution in common laboratory solvents, such as DMSO.
  • Characterized molecular weight and formula support reproducible experimental design.
  • Available in small research quantities appropriate for dose-response and in vivo studies.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-40704 Supplier Medchemexpress LLC Supplier No. HY1066285MG
Sudoxicam is a reversible, orally active cyclooxygenase (COX) antagonist from the enol-carboxamide class of non-steroidal anti-inflammatory drugs (NSAIDs). Provided for laboratory research use only, it exhibits anti-inflammatory, anti-edema, and antipyretic activities and is supplied as a characterized solid for use in in vitro and in vivo pharmacology studies.
  • Reversible COX antagonist with anti-inflammatory, anti-edema, and antipyretic activity.
  • Enol-carboxamide class NSAID commonly used in pharmacology research.
  • Supplied as a solid suitable for dissolution in common laboratory solvents, such as DMSO.
  • Characterized molecular weight and formula support reproducible experimental design.
  • Available in small research quantities appropriate for dose-response and in vivo studies.
  • Intended for research use only; not for human or clinical use.