Learn More
Medchemexpress LLC HY-W010254 500mg Medchemexpress, 4′-Hydroxy-2′-methylacetophenone CAS:875-59-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW010254500MG
Encompass_Preferred
Medchemexpress, HY-W010254 500mg 4′-Hydroxy-2′-methylacetophenone CAS:875-59-2 4′-Hydroxy-2′-methylacetophenone, an aroma compound of red wines, is isolated from cv. Bobal grape variety. 4′-Hydroxy-2′-methylacetophenone has ciliate toxicity. 4′-Hydroxy-2′-methylacetophenone inhibits the growth of T. pyriformis, with an IC50 of 0.65 mM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3287