Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxy-2-methylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. H14955G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H1495-5G 5 g
H14951G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H1495-5G Supplier TCI America Supplier No. H14955G
Add to Cart
Add to Cart

Specifications

CAS 493035-82-8
Melting Point 104°C
Color White-Yellow
Molecular Formula C7H9BO3
MDL Number MFCD03788424
Quantity 5 g
Synonym 4-hydroxy-2-methylbenzeneboronic acid, 4-hydroxy-2-methyl phenylboronic acid, 4-borono-3-methylphenol, 4-hydroxy-2-methylphenyl boronic acid, 4-hydroxy-2-methyl phenyl boronic acid
InChI Key OYIYNIONWDBJIF-UHFFFAOYSA-N
SMILES B(C1=C(C=C(C=C1)O)C)(O)O
IUPAC Name (4-hydroxy-2-methylphenyl)boronic acid
Molecular Weight (g/mol) 151.956
PubChem CID 2773448
Formula Weight 151.96
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.