Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxy-2'-nitrobiphenyl 97.0+%, TCI America™
SDP

Catalog No. H07531G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H07531G 1 g
H07535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H07531G Supplier TCI America Supplier No. H07531G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 51264-59-6
Molecular Formula C12H9NO3
Molecular Weight (g/mol) 215.208
MDL Number MFCD00143326
InChI Key NRRCMTITSCFYOH-UHFFFAOYSA-N
PubChem CID 15541112
IUPAC Name 4-(2-nitrophenyl)phenol
SMILES C1=CC=C(C(=C1)C2=CC=C(C=C2)O)[N+](=O)[O-]

Specifications

Melting Point 115°C
Quantity 1 g
Synonym 2′C-Nitrobiphenyl-4-ol, 4-(2-Nitrophenyl)phenol
Formula Weight 215.21
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-2′-nitrobiphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.