Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxy-3,5-dimethoxybenzyl alcohol, 97%

Catalog No. AAA1388814
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1388814 25 g
AAA1388803 1 g
AAA1388806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1388814 Supplier Thermo Scientific Chemicals Supplier No. A1388814
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 530-56-3 | C9H12O4 | 184.191 g/mol

4-Hydroxy-3,5-dimethoxybenzyl alcohol is a 2,4,6- substituted phenol with antiviral properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxy-3,5-dimethoxybenzyl alcohol is a 2,4,6- substituted phenol with antiviral properties.

Solubility
Sparingly soluble (0.40 g/L at 25°C).

Notes
Store in a cool, dry, well-ventilated area. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 530-56-3
Molecular Formula C9H12O4
Molecular Weight (g/mol) 184.191
MDL Number MFCD00016871
InChI Key LUOAEJWSKPQLJD-UHFFFAOYSA-N
Synonym 4-hydroxy-3,5-dimethoxybenzyl alcohol, 4-hydroxymethyl-2,6-dimethoxyphenol, syringyl alcohol, syringic alcohol, 3,5-dimethoxy-4-hydroxybenzyl alcohol, unii-k7g66i59xc, benzenemethanol, 4-hydroxy-3,5-dimethoxy, 2,6-dimethoxy-4-hydroxymethyl phenol, 4-hydroxymethyl-2,6-dimethoxy-phenol, 3,5-dimethoxy-4-hydroxybenzylalcohol
PubChem CID 10741
ChEBI CHEBI:86948
IUPAC Name 4-(hydroxymethyl)-2,6-dimethoxyphenol
SMILES COC1=CC(=CC(=C1O)OC)CO

Specifications

Melting Point 129°C to 132°C
Quantity 25 g
Beilstein 1873975
Solubility Information Sparingly soluble (0.40g/L at 25°C).
Formula Weight 184.19
Percent Purity 97%
Chemical Name or Material 4-Hydroxy-3,5-dimethoxybenzyl alcohol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 208-485-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.