Learn More
Medchemexpress LLC 4-Hydroxy-3-methoxy-5-nitrobenzoic acid | 15785-54-3 | 99.9% | C8H7NO6 | 10 G
SDP
Catalog No. 5000434870
Encompass_Preferred
4-Hydroxy-3-methoxy-5-nitrobenzoic acid is a drug intermediate used for the synthesis of various active compounds. This product is provided as a solid with high purity, intended for research applications only.
  • Used as a drug intermediate
  • Suitable for synthesis of active compounds
  • Supplied as a solid
  • High purity (99.9%)
  • Requires storage at 4°C under nitrogen
  • For research use only
Catalog No. 50-004-34870 Supplier Medchemexpress LLC Supplier No. HY7901210G
4-Hydroxy-3-methoxy-5-nitrobenzoic acid is a drug intermediate used for the synthesis of various active compounds. This product is provided as a solid with high purity, intended for research applications only.
  • Used as a drug intermediate
  • Suitable for synthesis of active compounds
  • Supplied as a solid
  • High purity (99.9%)
  • Requires storage at 4°C under nitrogen
  • For research use only