Learn More
Medchemexpress LLC 4-Hydroxy-3-methoxy-5-nitrobenzoic acid | 15785-54-3 | 99.9% | C8H7NO6 | 100 G
SDP
Catalog No. 5000435068
Encompass_Preferred
4-Hydroxy-3-methoxy-5-nitrobenzoic acid is a chemical compound that serves as a drug intermediate for the synthesis of various active compounds. This product is for research use only.
  • Used for synthesis of various active compounds
  • For research use only
  • Solid appearance
Catalog No. 50-004-35068 Supplier Medchemexpress LLC Supplier No. HY79012100G
4-Hydroxy-3-methoxy-5-nitrobenzoic acid is a chemical compound that serves as a drug intermediate for the synthesis of various active compounds. This product is for research use only.
  • Used for synthesis of various active compounds
  • For research use only
  • Solid appearance