Learn More
Medchemexpress LLC 4-Hydroxy-3-methoxy-5-nitrobenzoic acid | 15785-54-3 | 99.9% | C8H7NO6 | 25 G
SDP
Catalog No. 5000357561
Encompass_Preferred
4-Hydroxy-3-methoxy-5-nitrobenzoic acid, also known as 3-Nitro-4-hydroxy-5-methoxybenzoic acid, is a chemical compound that serves as a valuable drug intermediate in the synthesis of various active compounds. It is characterized by its high purity, as confirmed by its 99.94% assay.
  • Acts as a versatile drug intermediate.
  • Suitable for the synthesis of a wide range of active compounds.
  • High purity, with a typical assay of 99.94%.
Catalog No. 50-003-57561 Supplier Medchemexpress LLC Supplier No. 25GHY79012
Add to Cart
Add to Cart
4-Hydroxy-3-methoxy-5-nitrobenzoic acid, also known as 3-Nitro-4-hydroxy-5-methoxybenzoic acid, is a chemical compound that serves as a valuable drug intermediate in the synthesis of various active compounds. It is characterized by its high purity, as confirmed by its 99.94% assay.
  • Acts as a versatile drug intermediate.
  • Suitable for the synthesis of a wide range of active compounds.
  • High purity, with a typical assay of 99.94%.