Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-Hydroxy-3-methoxy-5-nitrobenzoic acid | 15785-54-3 | 99.9% | C8H7NO6 | 5 G
SDP

Catalog No. 5000434899
Encompass_Preferred

4-Hydroxy-3-methoxy-5-nitrobenzoic acid, also known as 3-Nitro-4-hydroxy-5-methoxybenzoic acid or 5-Nitrovanillic acid, is a drug intermediate employed in the synthesis of various active compounds. It appears as a light yellow to yellow solid with a molecular weight of 213.15. This product is intended for research use only.

  • Drug intermediate
  • Used in the synthesis of various active compounds
  • Solid appearance
  • Color: light yellow to yellow
  • Molecular weight: 213.15
  • For research use only

Catalog No. 50-004-34899 Supplier Medchemexpress LLC Supplier No. HY790125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Hydroxy-3-methoxy-5-nitrobenzoic acid, also known as 3-Nitro-4-hydroxy-5-methoxybenzoic acid or 5-Nitrovanillic acid, is a drug intermediate employed in the synthesis of various active compounds. It appears as a light yellow to yellow solid with a molecular weight of 213.15. This product is intended for research use only.

  • Drug intermediate
  • Used in the synthesis of various active compounds
  • Solid appearance
  • Color: light yellow to yellow
  • Molecular weight: 213.15
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.