Learn More
Medchemexpress LLC 4-hydroxy-3-methoxycinnamaldehyde | 458-36-6 | MFCD00075811 | 99.9% | 178.18 g/mol | C10H10O3 | 10 MG
SDP
Catalog No. 5000289397
Encompass_Preferred
Coniferaldehyde (4-hydroxy-3-methoxycinnamaldehyde) is a small-molecule research reagent reported to induce heme oxygenase-1 (HO-1) and to exhibit antioxidant and anti-inflammatory activity. It is supplied as a solid for analytical and laboratory applications and is intended for use in biochemical and cell-based studies.
  • Induces heme oxygenase-1 (HO-1) in cellular assays.
  • Demonstrates antioxidant and anti-inflammatory activity in preclinical studies.
  • Available in milligram quantities suitable for biochemical and cell-based experiments.
  • High assay purity for reproducible results.
  • Includes COA and SDS for quality and safety verification.
Catalog No. 50-002-89397 Supplier Medchemexpress LLC Supplier No. HYN253510MG
Add to Cart
Add to Cart
Coniferaldehyde (4-hydroxy-3-methoxycinnamaldehyde) is a small-molecule research reagent reported to induce heme oxygenase-1 (HO-1) and to exhibit antioxidant and anti-inflammatory activity. It is supplied as a solid for analytical and laboratory applications and is intended for use in biochemical and cell-based studies.
  • Induces heme oxygenase-1 (HO-1) in cellular assays.
  • Demonstrates antioxidant and anti-inflammatory activity in preclinical studies.
  • Available in milligram quantities suitable for biochemical and cell-based experiments.
  • High assay purity for reproducible results.
  • Includes COA and SDS for quality and safety verification.