Learn More
Medchemexpress LLC 4-hydroxy-3-methoxyphenylglycol sulfate (potassium) | 71324-20-4 | MFCD00043417 | 98.8% | 302.34 g/mol | C9H11KO7S | 5 MG
SDP
Catalog No. 5000264179
Encompass_Preferred
4-Hydroxy-3-methoxyphenylglycol sulfate potassium is the potassium salt of 3-methoxy-4-hydroxyphenylglycol sulfate supplied as a research-grade analytical reagent for studies of noradrenergic metabolism and related biochemical assays. It is a white to off-white solid with molecular formula C9H11KO7S, molecular weight 302.34 g/mol, and CAS 71324-20-4.
Catalog No. 50-002-64179 Supplier Medchemexpress LLC Supplier No. HY1134415MG
4-Hydroxy-3-methoxyphenylglycol sulfate potassium is the potassium salt of 3-methoxy-4-hydroxyphenylglycol sulfate supplied as a research-grade analytical reagent for studies of noradrenergic metabolism and related biochemical assays. It is a white to off-white solid with molecular formula C9H11KO7S, molecular weight 302.34 g/mol, and CAS 71324-20-4.