Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxy-3-methyl-2-butanone (Tech.), 85%, Thermo Scientific™

Catalog No. AC156371000
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC156371000 100 g
AC156370250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC156371000 Supplier Thermo Scientific Chemicals Supplier No. 156371000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3393-64-4
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.133
InChI Key VVSRECWZBBJOTG-UHFFFAOYSA-N
Synonym 4-hydroxy-3-methyl-2-butanone, 2-butanone, 4-hydroxy-3-methyl, 3-hydroxymethyl-2-butanone, 2-butanone, 4-hydroxy-3-methyl-, +, acmc-1cngd, 3-methyl-4-hydroxy-2-butanone, vvsrecwzbbjotg-uhfffaoysa, 2-butanone,4-hydroxy-3-methyl, 4-hydroxy-3-methyl-2-butanone, technical grade
PubChem CID 18829
IUPAC Name 4-hydroxy-3-methylbutan-2-one
SMILES CC(CO)C(=O)C

Specifications

CAS Min % 80.0
CAS Max % 100.0
Color Colorless
Density 0.99
Infrared Spectrum Authentic
Refractive Index 1.4320 to 1.4380 (20°C, 589nm)
Linear Formula HOCH2CH(CH3)COCH3
Quantity 100 g
Beilstein 01, I, 422
Formula Weight 102.13
Percent Purity ≥80%
Grade Technical
Physical Form Liquid
Chemical Name or Material 4-Hydroxy-3-methyl-2-butanone, tech., 85%
Show More Show Less
EINECSNumber 222-238-6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.