Learn More
Medchemexpress LLC 4'-Hydroxy-3'-methylacetophenone | 876-02-8 | MFCD00002231 | 150.18 | 500 MG
SDP
Catalog No. 5000447006
Encompass_Preferred
4'-Hydroxy-3'-methylacetophenone is a phenolic volatile compound that can be isolated from Hawaiian green coffee beans (Coffea Arabica L.). It is believed to possess potent antioxidant activity, similar to other phenolic volatile compounds like Thymol and Eugenol. This compound can also be used to synthesize heterocyclic compounds which have antimycobacterial activity.
  • Phenolic volatile compound
  • Isolated from Hawaiian green coffee beans (Coffea Arabica L.)
  • Possesses potent antioxidant activity
  • Used to synthesize heterocyclic compounds with antimycobacterial activity
Catalog No. 50-004-47006 Supplier Medchemexpress LLC Supplier No. HYW001663500MG
4'-Hydroxy-3'-methylacetophenone is a phenolic volatile compound that can be isolated from Hawaiian green coffee beans (Coffea Arabica L.). It is believed to possess potent antioxidant activity, similar to other phenolic volatile compounds like Thymol and Eugenol. This compound can also be used to synthesize heterocyclic compounds which have antimycobacterial activity.
  • Phenolic volatile compound
  • Isolated from Hawaiian green coffee beans (Coffea Arabica L.)
  • Possesses potent antioxidant activity
  • Used to synthesize heterocyclic compounds with antimycobacterial activity