Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxy-3-nitrobenzenearsonic Acid 98%, Thermo Scientific™

Catalog No. AC121665000
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC121665000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC121665000 Supplier Thermo Scientific Chemicals Supplier No. 121665000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-19-7
Molecular Formula C6H6AsNO6
Molecular Weight (g/mol) 263.037
MDL Number MFCD00007112
InChI Key XMVJITFPVVRMHC-UHFFFAOYSA-N
Synonym roxarsone, 3-nitro, ristat, 4-hydroxy-3-nitrobenzenearsonic acid, aklomix-3, roxarson, ren-o-sal, 3-nitro-10, 3n4hpa, nitrophenolarsonic acid
PubChem CID 5104
ChEBI CHEBI:35817
IUPAC Name (4-hydroxy-3-nitrophenyl)arsonic acid
SMILES C1=CC(=C(C=C1[As](=O)(O)O)[N+](=O)[O-])O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Beige-Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula HOC6H3(NO2)AsO3H2
Quantity 500 g
Beilstein 16, I, 456
Merck Index 15, 8406
Formula Weight 263.03
Percent Purity ≥97.5% (on dry substance)
Physical Form Fine Crystalline Powder
Chemical Name or Material 4-Hydroxy-3-nitrobenzenearsonic acid, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Toxic if inhaled.
Precautionary Statement GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 204-453-7
RTECSNumber CY5250000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.