Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxy-3-nitrobiphenyl, TCI America™
SDP

Catalog No. N056025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N056025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N056025G Supplier TCI America Supplier No. N056025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 885-82-5
Molecular Formula C12H9NO3
Molecular Weight (g/mol) 215.208
MDL Number MFCD00014694
InChI Key JDDNJJBXFOLPKX-UHFFFAOYSA-N
Synonym 4-hydroxy-3-nitrobiphenyl, 3-nitrobiphenyl-4-ol, 3-nitro-1,1'-biphenyl-4-ol, 3-nitro 1,1'-biphenyl-4-ol, ccris 5769, 2-nitro-4-phenyl-phenol, 4-phenyl-2-nitrophenol, acmc-209qum, nitrophenylphenol, 3-nitro-4-hydroxybiphenyl
PubChem CID 13447
IUPAC Name 2-nitro-4-phenylphenol
SMILES C1=CC=C(C=C1)C2=CC(=C(C=C2)O)[N+](=O)[O-]

Specifications

Melting Point 69°C
Quantity 25 g
Formula Weight 215.21
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-3-nitrobiphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.