Learn More
Sigma Aldrich 4-Hydroxy-4-methyl-2-pentanone
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- 4-hydroxy-4-methyl-2-pentanone can be used to synthesize:
- Diolmonoesters by reacting with aldehydes via tandem aldol-transfer-tischtschenko reaction catalyzed by trimethylaluminum
- Aldol-transfer products such as α-hydroxy ketones by reacting with various aldehydes in the presence of L-proline as a catalyst
- Mono-α-arylated ketones and esters via pd catalyzed reaction with aryl halides and aryl triflates, respectively.it can also be used as a complexing agent to synthesize double-metal cyanide (dmc) catalysts by reacting with metal salt (mx) and cyanometalate compounds
Specifications
Specifications
| CAS | 123-42-2 |
| Density | 0.931 g/mL (at 25°C (literature)) |
| Boiling Point | 166°C (lit.) |
| Molecular Formula | C6H12O2 |
| Refractive Index | n20/D 1.423 (literature) |
| Linear Formula | (CH3)2 C(OH)CH2COCH3 |
| MDL Number | MFCD00004471 |
| Quantity | 25 mL |
| Synonym | Diacetone alcohol |
| Molecular Weight (g/mol) | 116.16 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.