Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Hydroxy-4-methyl-2-pentanone

Catalog No. H41544500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
500 mL
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H41544500ML 500 mL
H415444L 4 L
H4154425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H41544500ML Supplier Sigma Aldrich Supplier No. H41544500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 4-hydroxy-4-methyl-2-pentanone can be used to synthesize:
  • Diolmonoesters by reacting with aldehydes via tandem aldol-transfer-tischtschenko reaction catalyzed by trimethylaluminum
  • Aldol-transfer products such as α-hydroxy ketones by reacting with various aldehydes in the presence of L-proline as a catalyst
  • Mono-α-arylated ketones and esters via pd catalyzed reaction with aryl halides and aryl triflates, respectively.it can also be used as a complexing agent to synthesize double-metal cyanide (dmc) catalysts by reacting with metal salt (mx) and cyanometalate compounds

Specifications

CAS 123-42-2
Density 0.931 g/mL (at 25°C (literature))
Boiling Point 166°C (lit.)
Molecular Formula C6H12O2
Linear Formula (CH3)2 C(OH)CH2COCH3
Refractive Index n20/D 1.423 (literature)
MDL Number MFCD00004471
Quantity 500 mL
Synonym Diacetone alcohol
Molecular Weight (g/mol) 116.16
Percent Purity 99%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.