Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxy-4-methyl-2-pentanone, 98+%

Catalog No. AAA162480F
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA162480F 2500 mL
AAA16248AE 100 mL
AAA16248AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA162480F Supplier Thermo Scientific Chemicals Supplier No. A162480F
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 123-42-2 | C6H12O2 | 116.16 g/mol

4-Hydroxy-4-methyl-2-pentanone, is used as a solvent. It is also used as an intermediate of chemical products and is used as solvent for some industrial paint to adjust thesolubility of paint resins and regulate the evaporation rate. It is mainly used in paints and coatings (coil coatings, wood varnishes, architectural coatings…). It is a good solvent for organic peroxides.It can also be used for the treatment of textiles and leather, in chemical synthesis or as a cleaning solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxy-4-methyl-2-pentanone, is used as a solvent. It is also used as an intermediate of chemical products and is used as solvent for some industrial paint to adjust thesolubility of paint resins and regulate the evaporation rate. It is mainly used in paints and coatings (coil coatings, wood varnishes, architectural coatings…). It is a good solvent for organic peroxides.It can also be used for the treatment of textiles and leather, in chemical synthesis or as a cleaning solvent.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-42-2
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00004471
InChI Key SWXVUIWOUIDPGS-UHFFFAOYSA-N
Synonym diacetone alcohol, 4-hydroxy-4-methyl-2-pentanone, diacetonalkohol, diketone alcohol, diacetonalcohol, tyranton, acetonyldimethylcarbinol, diacetone-alcool, pyranton a, 2-methyl-2-pentanol-4-one
PubChem CID 31256
ChEBI CHEBI:55381
IUPAC Name 4-hydroxy-4-methylpentan-2-one
SMILES CC(=O)CC(C)(C)O

Specifications

Melting Point -47°C
Density 0.935
Boiling Point 168°C
Flash Point 58°C (136°F)
Odor Pleasant
Refractive Index 1.423
Quantity 2500 mL
UN Number UN1148
Beilstein 1740440
Merck Index 14,2964
Solubility Information It is soluble in water.
Formula Weight 116.16
Percent Purity ≥98%
Chemical Name or Material 4-Hydroxy-4-methyl-2-pentanone
Show More Show Less
Hazard Category H226-H319-H335
Hazard Statement GHS H Statement
H226-H319
Flammable liquid and vapour.
Causes serious eye irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: DIACETONE ALCOHOL
EINECSNumber 204-626-7
RTECSNumber SA9100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.