Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxy-6-methyl-3-nitro-2-pyridone, 97%

Catalog No. AAA1969022
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1969022 100 g
AAA1969014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1969022 Supplier Thermo Scientific Chemicals Supplier No. A1969022
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 4966-90-9 | C6H6N2O4 | 170.12 g/mol

It is employed as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a pharmaceutical intermediate.

Solubility
Solubility in N,N-DMF gives very faint turbidity.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4966-90-9
Molecular Formula C6H6N2O4
Molecular Weight (g/mol) 170.12
MDL Number MFCD00167612
InChI Key QIKWTNPFTOEELW-UHFFFAOYSA-N
Synonym 4-hydroxy-6-methyl-3-nitro-2-pyridone, 4-hydroxy-6-methyl-3-nitropyridin-2 1h-one, 6-methyl-3-nitropyridine-2,4-diol, 2,4-dihydroxy-6-methyl-3-nitropyridine, 2-hydroxy-6-methyl-3-nitro-1h-pyridin-4-one, 4-hydroxy-6-methyl-3-nitro-1,2-dihydropyridin-2-one, 4-hydroxy-6-methyl-3-nitro-2 1h-pyridinone, 4-hydroxy-6-methyl-3-nitrohydropyridin-2-one, pubchem3892, acmc-209khe
PubChem CID 54685619
IUPAC Name 4-hydroxy-6-methyl-3-nitro-1H-pyridin-2-one
SMILES CC1=CC(=O)C(=C(O)N1)[N+]([O-])=O

Specifications

Melting Point >250°C (decomposition)
Quantity 100 g
Beilstein 1529425
Formula Weight 170.13
Percent Purity 97%
Chemical Name or Material 4-Hydroxy-6-methyl-3-nitro-2-pyridone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.