Learn More
Cayman Chemical 4-hydroxy Atorvastatin calci
SDP
Catalog No. 502964097
Encompass_Preferred
A metabolite of atorvastatin; formed by metabolism of atorvastatin by CYP3A4
Catalog No. 50-296-4097 Supplier Cayman Chemical Supplier No. 22575500 UG
A metabolite of atorvastatin; formed by metabolism of atorvastatin by CYP3A4