Learn More
Cayman Chemical 4-hydroxy DALT, Purity: ≥98%, CAS: 2173386-70-2, 1 mg
SDP
Catalog No. 502912262
Encompass_Preferred
An analytical reference standard categorized as a tryptamine; induces the HTR in mice; intended for research and forensic applications
Catalog No. 50-291-2262 Supplier Cayman Chemical Supplier No. 347891 MG
An analytical reference standard categorized as a tryptamine; induces the HTR in mice; intended for research and forensic applications