Learn More
Cayman Chemical 4-hydroxy Estrone, Purity: ≥95%, CAS: 3131-23-5, 10 mg
SDP
Catalog No. 502947044
Encompass_Preferred
A metabolite of estrone; induces nuclear translocation of phosphorylated p53 and is protective against glutamate-induced oxidative death in HT22 cells at 5 µM; reduces kainic acid-induced hippocampal neurodegeneration and improves working memory in rats at 10 µg/animal; also inhibits proliferation of MCF-7 and MDA-MB-231 breast cancer cells (IC50s = 30 and 38 µM, respectively)
Catalog No. 50-294-7044 Supplier Cayman Chemical Supplier No. 3369610 MG
A metabolite of estrone; induces nuclear translocation of phosphorylated p53 and is protective against glutamate-induced oxidative death in HT22 cells at 5 µM; reduces kainic acid-induced hippocampal neurodegeneration and improves working memory in rats at 10 µg/animal; also inhibits proliferation of MCF-7 and MDA-MB-231 breast cancer cells (IC50s = 30 and 38 µM, respectively)