Learn More
Cayman Chemical 4'-hydroxy Flurbiprofen, Purity: ≥95%, CAS: 52807-12-2, 10 mg
SDP
Catalog No. 502985860
Encompass_Preferred
A major active metabolite of flurbiprofen, (R)-flurbiprofen, and (S)-flurbiprofen; inhibits COX-1 by 94% at 1,000 µM but does not inhibit oxygenation of arachidonic acid; completely inhibits cyclooxygenation of 2-AG at 300 µM; inhibits FAAH hydrolysis of anandamide in rat brain homogenates (IC50 = 84 µM at a pH of 6)
Catalog No. 50-298-5860 Supplier Cayman Chemical Supplier No. 2169110 MG
A major active metabolite of flurbiprofen, (R)-flurbiprofen, and (S)-flurbiprofen; inhibits COX-1 by 94% at 1,000 µM but does not inhibit oxygenation of arachidonic acid; completely inhibits cyclooxygenation of 2-AG at 300 µM; inhibits FAAH hydrolysis of anandamide in rat brain homogenates (IC50 = 84 µM at a pH of 6)