Learn More
Cayman Chemical 4-hydroxy Hexenal, Purity: ≥98%, CAS: 17427-21-3, 25 mg
SDP
Catalog No. 502946896
Encompass_Preferred
A lipid peroxidation product derived from oxidized ω-3 fatty acids such as DHA
Catalog No. 50-294-6896 Supplier Cayman Chemical Supplier No. 3206025 MG
A lipid peroxidation product derived from oxidized ω-3 fatty acids such as DHA