Learn More
Medchemexpress LLC 4-hydroxy-L-proline methyl ester hydrochloride | 40216-83-9 | MFCD00080855 | >97.0% | 181.62 | C6H12ClNO3 | 1 KG
SDP
Catalog No. 5000203501
Encompass_Preferred
H-Hyp-OMe hydrochloride is the methyl ester hydrochloride of trans-4-hydroxy-L-proline, used as a building block and non-cleavable linker in organic synthesis and bioconjugation.
  • Used as a linker or building block in antibody-drug conjugate and PROTAC synthesis.
  • Methyl ester hydrochloride form facilitates coupling reactions in organic synthesis.
  • Available in bulk quantities suitable for scale-up reactions.
  • Stable when stored sealed at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Reported purity meets typical requirements for synthetic applications.
Catalog No. 50-002-03501 Supplier Medchemexpress LLC Supplier No. HY760431KG
H-Hyp-OMe hydrochloride is the methyl ester hydrochloride of trans-4-hydroxy-L-proline, used as a building block and non-cleavable linker in organic synthesis and bioconjugation.
  • Used as a linker or building block in antibody-drug conjugate and PROTAC synthesis.
  • Methyl ester hydrochloride form facilitates coupling reactions in organic synthesis.
  • Available in bulk quantities suitable for scale-up reactions.
  • Stable when stored sealed at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Reported purity meets typical requirements for synthetic applications.