Learn More
Cayman Chemical 4-hydroxy MET, Purity: ≥98%, CAS: 77872-41-4, 5 mg
SDP
Catalog No. 502953456
Encompass_Preferred
An analytical reference standard categorized as a tryptamine; induces the HTR in mice; intended for research and forensic applications
Catalog No. 50-295-3456 Supplier Cayman Chemical Supplier No. 111485 MG
An analytical reference standard categorized as a tryptamine; induces the HTR in mice; intended for research and forensic applications