Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide hydrochloride | 2115897-23-7 | MFCD32062443 | 99.2% | 467.02 | C22H31ClN4O3S | 10MG
SDP

Catalog No. 5000239518
Encompass_Preferred

(S,S,S)-AHPC hydrochloride is a research-grade VHL ligand provided as the hydrochloride salt. It is used as a building block or negative control in protein degrader (PROTAC) development and biochemical studies of von Hippel-Lindau (VHL) E3 ligase recruitment.

  • Research-grade VHL ligand for protein degrader and binding studies.
  • High purity suitable for biochemical and cell-based assays (99.2%).
  • Hydrochloride salt form to enhance solubility in polar solvents.
  • Molecular weight 467.02 and formula C22H31ClN4O3S.
  • Supplied as a 10 mg vial for small-scale research use.

Catalog No. 50-002-39518 Supplier Medchemexpress LLC Supplier No. HY125845A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S,S,S)-AHPC hydrochloride is a research-grade VHL ligand provided as the hydrochloride salt. It is used as a building block or negative control in protein degrader (PROTAC) development and biochemical studies of von Hippel-Lindau (VHL) E3 ligase recruitment.

  • Research-grade VHL ligand for protein degrader and binding studies.
  • High purity suitable for biochemical and cell-based assays (99.2%).
  • Hydrochloride salt form to enhance solubility in polar solvents.
  • Molecular weight 467.02 and formula C22H31ClN4O3S.
  • Supplied as a 10 mg vial for small-scale research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.