Learn More
Cayman Chemical 4-hydroxy Nebivolol hydrochl
SDP
Catalog No. 502932683
Encompass_Preferred
A major metabolite of nebivolol; formed by the hydroxylation of nebivolol by CYP2D6
Catalog No. 50-293-2683 Supplier Cayman Chemical Supplier No. 26658500 UG
A major metabolite of nebivolol; formed by the hydroxylation of nebivolol by CYP2D6