Learn More
Cayman Chemical 4-hydroxy Nonenal, Purity: ≥98%, CAS: 75899-68-2, 10 mg
SDP
Catalog No. 502947611
Encompass_Preferred
A lipid peroxidation product derived from oxidized ω-6 PUFAs such as linoleic acid and arachidonic acid which is widely used as a marker of lipid peroxidation
Catalog No. 50-294-7611 Supplier Cayman Chemical Supplier No. 3210010 MG
A lipid peroxidation product derived from oxidized ω-6 PUFAs such as linoleic acid and arachidonic acid which is widely used as a marker of lipid peroxidation