Learn More
Cayman Chemical 4-hydroxy Nonal-d3 100ug
SDP
Catalog No. 502946629
Encompass_Preferred
An internal standard for the quantification of 4-HNE by GC- or LC-MS
Catalog No. 50-294-6629 Supplier Cayman Chemical Supplier No. 332101100 UG
An internal standard for the quantification of 4-HNE by GC- or LC-MS