Learn More
Cayman Chemical 4-hydroxy Nonal-d3 1mg
SDP
Catalog No. 502947627
Encompass_Preferred
An internal standard for the quantification of 4-HNE by GC- or LC-MS
Catalog No. 50-294-7627 Supplier Cayman Chemical Supplier No. 3321011 MG
An internal standard for the quantification of 4-HNE by GC- or LC-MS